Assay Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015729.3 | NP_056544.2 | 3 - 4 | 768 | 59 | |
| GenBank mRNA | AK040566.1 | - | 3 - 4 | 471 | 59 | |
| AK054446.1 | - | 3 - 4 | 544 | 59 | ||
| BC054727.1 | - | 3 - 4 | 436 | 59 | ||
| BC056448.1 | - | 3 - 4 | 768 | 5704873 | 59 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

Acox1
Genomic Map