Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200017873.mm.1 |
NM_001289738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017873.mm.1 |
NM_177656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017873.mm.1 |
NR_110367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017873.mm.1 |
NR_110368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017873.mm.1 |
TC0200002282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017876.mm.1 |
NM_001289738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017876.mm.1 |
NM_177656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017876.mm.1 |
NR_110367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017876.mm.1 |
NR_110368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017876.mm.1 |
TC0200002282.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200017874.mm.1 |
NM_001289738.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017874.mm.1 |
NM_177656.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017874.mm.1 |
NR_110367.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017874.mm.1 |
NR_110368.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200017874.mm.1 |
TC0200002282.mm.1 |
MTA TC |