Assay Details
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178624.6 | NP_848739.1 | 11 - 12 | 905 | 58 | |
GenBank mRNA | AK013142.1 | - | 12 - 13 | 1039 | 58 | |
AK039010.1 | - | 11 - 12 | 905 | 58 | ||
AK045742.1 | - | 11 - 12 | 892 | 58 | ||
AK089994.1 | - | 11 - 12 | 881 | 58 | ||
AK141177.1 | - | 11 - 12 | 895 | 58 | ||
AK163472.1 | - | 11 - 12 | 901 | 58 | ||
BC051530.1 | - | 7 - 8 | 719 | 58 | ||
BC096582.1 | - | 11 - 12 | 891 | 3963599 | 58 | |
BC145665.1 | - | 11 - 12 | 885 | 40130968 | 58 | |
BC145998.1 | - | 11 - 12 | 885 | 40131301 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
