Assay Details
Gene Symbol: Fbxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178624.6 | NP_848739.1 | 1 - 2 | 58 | 92 | |
GenBank mRNA | AK039010.1 | - | 1 - 2 | 58 | 92 | |
AK044693.1 | - | 1 - 2 | 55 | 92 | ||
AK045742.1 | - | 1 - 2 | 45 | 92 | ||
AK141177.1 | - | 1 - 2 | 48 | 92 | ||
AK163472.1 | - | 1 - 2 | 54 | 92 | ||
BC096582.1 | - | 1 - 2 | 44 | 3963599 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK039010.1, AK044693.1, AK045742.1, AK141177.1, AK163472.1, BC096582.1, NM_178624.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
