Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081045.1 | NP_001074514.1 | 10 - 11 | 3116 | 108 | |
| GenBank mRNA | AK006970.1 | - | 2 - 3 | 227 | 108 | |
| AK122484.1 | - | 10 - 11 | 3122 | 108 | ||
| AK161032.1 | - | 1 - 2 | 151 | 108 | ||
| BC043121.1 | - | 10 - 11 | 2665 | 6409084 | 108 | |
| BC054752.1 | - | 11 - 12 | 2939 | 6834541 | 108 | |
| BC079594.1 | - | 10 - 11 | 2664 | 6827379 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1
Genomic Map