Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081045.1 | NP_001074514.1 | 3 - 4 | 2077 | 98 | |
| GenBank mRNA | AK037800.1 | - | 4 - 5 | 1643 | 98 | |
| AK122484.1 | - | 3 - 4 | 2083 | 98 | ||
| BC043121.1 | - | 3 - 4 | 1626 | 6409084 | 98 | |
| BC054752.1 | - | 4 - 5 | 1900 | 6834541 | 98 | |
| BC079594.1 | - | 3 - 4 | 1625 | 6827379 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1
Genomic Map