Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081045.1 | NP_001074514.1 | 4 - 5 | 2182 | 110 | |
| GenBank mRNA | AK037800.1 | - | 5 - 6 | 1748 | 110 | |
| AK122484.1 | - | 4 - 5 | 2188 | 110 | ||
| BC043121.1 | - | 4 - 5 | 1731 | 6409084 | 110 | |
| BC054752.1 | - | 5 - 6 | 2005 | 6834541 | 110 | |
| BC079594.1 | - | 4 - 5 | 1730 | 6827379 | 110 | |
More Information
Set Membership: |
|

Kansl1
Genomic Map