Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081045.1 | NP_001074514.1 | 6 - 7 | 2489 | 74 | |
| GenBank mRNA | AK122484.1 | - | 6 - 7 | 2495 | 74 | |
| BC043121.1 | - | 6 - 7 | 2038 | 6409084 | 74 | |
| BC054752.1 | - | 7 - 8 | 2312 | 6834541 | 74 | |
| BC079594.1 | - | 6 - 7 | 2037 | 6827379 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1
Genomic Map