Assay Details
| Gene Symbol: Kcnh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037712.1 | NP_001032801.1 | 10 - 11 | 2238 | 77 | |
| GenBank mRNA | AK038513.1 | - | 4 - 5 | 737 | 77 | |
| BC109174.1 | - | 10 - 11 | 2238 | 40048550 | 77 | |
| BC109175.1 | - | 10 - 11 | 2343 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh6
Genomic Map