Assay Details
Gene Symbol: Kcnh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037712.1 | NP_001032801.1 | 1 - 2 | 81 | 63 | |
GenBank mRNA | BC109174.1 | - | 1 - 2 | 81 | 40048550 | 63 |
BC109175.1 | - | 1 - 2 | 81 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
