Assay Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049832.1 | - | 1 - 2 | 119 | 89 | |
| AK146380.1 | - | 1 - 2 | 113 | 89 | ||
| AK150427.1 | - | 1 - 2 | 119 | 89 | ||
| AK151124.1 | - | 1 - 2 | 85 | 89 | ||
| AK153227.1 | - | 1 - 2 | 85 | 89 | ||
| AK158562.1 | - | 1 - 2 | 464 | 89 | ||
| BC005697.1 | - | 1 - 2 | 68 | 3597765 | 89 | |
| HQ412666.1 | - | 2 - 3 | 309 | 89 | ||
| HQ412668.1 | - | 1 - 2 | 185 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150427.1, AK151124.1, AK153227.1, BC005697.1, HQ412666.1, HQ412668.1 transcripts.
Set Membership: |
|

Prkar1a
Genomic Map