Assay Details
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291114.1 | NP_001278043.1 | 12 - 13 | 1538 | 58 | |
| NM_001291115.1 | NP_001278044.1 | 12 - 13 | 1333 | 58 | ||
| GenBank mRNA | AK134091.1 | - | 2 - 3 | 108 | 58 | |
| AK135289.1 | - | 11 - 12 | 1164 | 58 | ||
| AK146046.1 | - | 12 - 13 | 1333 | 58 | ||
| AK154989.1 | - | 12 - 13 | 1469 | 58 | ||
| BC030493.1 | - | 10 - 11 | 1153 | 5400675 | 58 | |
| BC082607.1 | - | 10 - 11 | 1128 | 30667785 | 58 | |
| BC092268.1 | - | 10 - 11 | 1287 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm39
Genomic Map