Assay Details
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291114.1 | NP_001278043.1 | 10 - 11 | 1253 | 78 | |
| NM_001291115.1 | NP_001278044.1 | 10 - 11 | 1048 | 78 | ||
| NM_133242.3 | NP_573505.2 | 10 - 11 | 1253 | 78 | ||
| GenBank mRNA | AK135289.1 | - | 9 - 10 | 879 | 78 | |
| AK147076.1 | - | 10 - 11 | 1198 | 78 | ||
| AK154989.1 | - | 10 - 11 | 1184 | 78 | ||
| BC004000.1 | - | 6 - 7 | 585 | 78 | ||
| BC023205.1 | - | 10 - 11 | 1155 | 78 | ||
| BC030493.1 | - | 8 - 9 | 868 | 5400675 | 78 | |
| BC082607.1 | - | 8 - 9 | 843 | 30667785 | 78 | |
| BC086645.1 | - | 10 - 11 | 1176 | 6817169 | 78 | |
| BC092268.1 | - | 8 - 9 | 1002 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm39
Genomic Map