Assay Details
| Gene Symbol: Zbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021394.2 | NP_067369.2 | 3 - 4 | 603 | 94 | |
| GenBank mRNA | AF136520.1 | - | 3 - 4 | 571 | 94 | |
| AK157215.1 | - | 3 - 4 | 579 | 94 | ||
| AX775969.1 | - | 3 - 4 | 557 | 94 | ||
| AX775971.1 | - | 3 - 4 | 557 | 94 | ||
| BC020033.1 | - | 3 - 4 | 573 | 4007636 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zbp1
Genomic Map