Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800019101.mm.1 |
NM_001191004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019101.mm.1 |
NM_001191004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019101.mm.1 |
NM_030145.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019101.mm.1 |
NM_030145.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019101.mm.1 |
TC0800002512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019103.mm.1 |
NM_001191004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019103.mm.1 |
NM_001191004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019103.mm.1 |
NM_030145.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019103.mm.1 |
NM_030145.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019103.mm.1 |
TC0800002512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019095.mm.1 |
NM_001191004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019095.mm.1 |
NM_001191004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019095.mm.1 |
NM_030145.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019095.mm.1 |
NM_030145.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019095.mm.1 |
TC0800002512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019097.mm.1 |
NM_001191004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019097.mm.1 |
NM_001191004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019097.mm.1 |
NM_030145.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019097.mm.1 |
NM_030145.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019097.mm.1 |
TC0800002512.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800019099.mm.1 |
NM_001191004.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019099.mm.1 |
NM_001191004.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800019099.mm.1 |
TC0800002512.mm.1 |
MTA TC |