Assay Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024617.3 | NP_001019788.1 | 16 - 17 | 1986 | 92 | |
| NM_001297591.1 | NP_001284520.1 | 16 - 17 | 1963 | 92 | ||
| NM_001297593.1 | NP_001284522.1 | 15 - 16 | 1844 | 92 | ||
| NM_001297596.1 | NP_001284525.1 | 16 - 17 | 1963 | 92 | ||
| GenBank mRNA | AK053554.1 | - | 9 - 10 | 1115 | 92 | |
| AK089068.1 | - | 11 - 12 | 1217 | 92 | ||
| AK139397.1 | - | 9 - 10 | 1463 | 92 | ||
| AK141502.1 | - | 18 - 19 | 1960 | 92 | ||
| BC044876.1 | - | 2 - 3 | 350 | 92 | ||
| BC064813.1 | - | 14 - 15 | 1688 | 6826283 | 92 | |
| DQ296611.1 | - | 16 - 17 | 1963 | 92 | ||
| DQ296612.1 | - | 15 - 16 | 1844 | 92 | ||
| DQ296613.1 | - | 16 - 17 | 1963 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Inpp4b
Genomic Map