Assay Details
| Gene Symbol: Inpp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297591.1 | NP_001284520.1 | 6 - 7 | 774 | 137 | |
| NM_001297593.1 | NP_001284522.1 | 5 - 6 | 655 | 137 | ||
| NM_001297596.1 | NP_001284525.1 | 6 - 7 | 774 | 137 | ||
| GenBank mRNA | AK141502.1 | - | 8 - 9 | 771 | 137 | |
| DQ296611.1 | - | 6 - 7 | 774 | 137 | ||
| DQ296612.1 | - | 5 - 6 | 655 | 137 | ||
| DQ296613.1 | - | 6 - 7 | 774 | 137 | ||
More Information
Set Membership: |
|

Inpp4b
Genomic Map