Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200020088.mm.1 |
NR_040617.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020088.mm.1 |
NR_040617.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020088.mm.1 |
TC0200002468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020090.mm.1 |
NR_040617.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020090.mm.1 |
NR_040617.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020090.mm.1 |
TC0200002468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020089.mm.1 |
NR_040617.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020089.mm.1 |
NR_040617.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020089.mm.1 |
TC0200002468.mm.1 |
MTA TC |