Assay Details
| Gene Symbol: Cdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009864.2 | NP_033994.1 | 12 - 13 | 2071 | 84 | |
| GenBank mRNA | AK076369.1 | - | 12 - 13 | 2073 | 84 | |
| AK166267.1 | - | 12 - 13 | 2073 | 84 | ||
| BC098501.1 | - | 12 - 13 | 2060 | 30023851 | 84 | |
| X06115.1 | - | 12 - 13 | 2012 | 84 | ||
| X06339.1 | - | 9 - 10 | 1425 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh1
Genomic Map