Assay Details
| Gene Symbol: Wwox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019573.3 | NP_062519.2 | 1 - 2 | 227 | 74 | |
| GenBank mRNA | AF187014.1 | - | 1 - 2 | 178 | 74 | |
| AK018507.1 | - | 1 - 2 | 157 | 74 | ||
| AK019911.1 | - | 1 - 2 | 185 | 74 | ||
| AK046903.1 | - | 1 - 2 | 177 | 74 | ||
| AK050541.1 | - | 1 - 2 | 178 | 74 | ||
| AK078528.1 | - | 1 - 2 | 180 | 74 | ||
| AK083096.1 | - | 1 - 2 | 159 | 74 | ||
| BC014716.1 | - | 1 - 2 | 183 | 4241066 | 74 | |
| CJ058964.1 | - | 1 - 2 | 228 | 74 | ||
More Information
Set Membership: |
|

Wwox
Genomic Map