Assay Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028532.3 | NP_082808.1 | 1 - 2 | 92 | 70 | |
| GenBank mRNA | AK006802.1 | - | 1 - 2 | 79 | 70 | |
| BC104382.1 | - | 1 - 2 | 91 | 70 | ||
| BC104383.1 | - | 1 - 2 | 92 | 70 | ||
| BY706785.1 | - | 1 - 2 | 77 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006802.1, BC104382.1, BC104383.1, NM_028532.3 transcripts.
Set Membership: |
|

Tepp
Genomic Map