Assay Details
Gene Symbol: Tepp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028532.3 | NP_082808.1 | 6 - 7 | 490 | 63 | |
NM_199455.2 | NP_955534.1 | 7 - 8 | 715 | 63 | ||
GenBank mRNA | AK006802.1 | - | 6 - 7 | 477 | 63 | |
BC087870.1 | - | 7 - 8 | 711 | 6707505 | 63 | |
BC104382.1 | - | 6 - 7 | 489 | 63 | ||
BC104383.1 | - | 6 - 7 | 490 | 63 | ||
BY706785.1 | - | 6 - 7 | 475 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082618 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082616 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082617 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082619 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | BC104382.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | BC104383.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | NM_028532.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | NM_199455.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | AK006802.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | BC104382.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | BC104383.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | NM_028532.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | NM_199455.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P450682 | AK006802.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407671 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 142686 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119568 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | BC104382.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | BC104383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | NM_028532.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | NM_199455.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | AK006802.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | BC104382.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | BC104383.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | NM_028532.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | NM_199455.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P450682 | AK006802.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082590 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082593 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082586 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082587 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082589 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082591 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26082588 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06605027 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06605036 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119568 | - | - |
Whole Mouse Genome G4122A | A_51_P450682 | BC104382.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | BC104383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | NM_028532.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | NM_199455.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | AK006802.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | BC104382.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | BC104383.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | NM_028532.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | NM_199455.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P450682 | AK006802.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125682 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109001 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119568 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06605027 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06964775 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06964774 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082619 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082618 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082616 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26082617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082603 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082622 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082703 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082653 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26082631 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082599 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06777878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082604 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07241139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06998908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082596 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26082603 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10574340 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574332 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574341 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574331 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574338 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574339 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574334 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574336 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574333 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574335 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432536_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574330 | - | - |
Mouse Genome 430 2.0 Array | 1432536_at | - | - |
Mouse Genome 430 2.0 Array | 1432536_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73407_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73407_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800008908.mm.1 | NM_028532.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008908.mm.1 | NM_199455.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008908.mm.1 | TC0800001131.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432536_at | - | - |
Mouse Expression Set 430 | 1432536_PM_at | - | - |
Murine Genome U74v2 Set | 170181_at | - | - |
Murine Genome U74v2 Set | 169867_r_at | - | - |
Murine Genome U74v2 Set | 170181_PM_at | - | - |
Murine Genome U74v2 Set | 169867_PM_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432536_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432536_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1450687 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1780167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001780167 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001450687 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1226573 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2606189 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1226573 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2606189 | - | - |
mouse-6 v1.1 (Illumina) | scl073407.3_30-S | - | - |
mouse-6 v1.1 (Illumina) | scl000721.1_129-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1780167 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1450687 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl073407.3_30-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl000721.1_129-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1450687 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780167 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1226573 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2606189 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2606189 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1226573 | - | - |
