Assay Details
Gene Symbol: 4931428F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166394.1 | NP_001159866.1 | 6 - 7 | 1420 | 71 | |
NM_028888.2 | NP_083164.1 | 7 - 8 | 1817 | 71 | ||
GenBank mRNA | AK016481.1 | - | 6 - 7 | 1755 | 71 | |
AK136929.1 | - | 6 - 7 | 1582 | 71 | ||
BC043046.1 | - | 6 - 7 | 1577 | 5703896 | 71 | |
BC059045.1 | - | 6 - 7 | 1355 | 6816353 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132761 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132762 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132763 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132764 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132765 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | AK053846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | AK079536.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | AK197583.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | NM_030152.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | BC043046.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | AK053846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | AK079536.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | AK197583.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | NM_030152.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P222096 | BC043046.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135045 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358331 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 251114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782307 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519729 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 171554 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85604 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113722 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109176 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153034 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | AK053846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | AK079536.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | AK197583.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | NM_030152.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | BC043046.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | AK053846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | AK079536.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | AK197583.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | NM_030152.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P222096 | BC043046.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132755 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132757 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26132763 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153034 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109176 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113722 | - | - |
Whole Mouse Genome G4122A | A_52_P222096 | AK053846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | AK079536.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | AK197583.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | NM_030152.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | BC043046.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | AK053846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | AK079536.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | AK197583.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | NM_030152.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P222096 | BC043046.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P105241 | AK016481.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P620602 | CJ064766.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P620602 | AK049007.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P620602 | NM_028888.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108890 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153034 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109176 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113722 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390409 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390399 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07390412 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132764 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132762 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132763 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132765 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26132761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132686 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132682 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132691 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132655 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132678 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132680 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26132743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06922511 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132716 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07255497 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132749 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132755 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07255503 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06922519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132728 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26132745 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581192 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581196 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581194 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581190 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581195 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581189 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581191 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581193 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1440743_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1435024_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429756_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574676 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581188 | - | - |
Mouse Genome 430 2.0 Array | 1429756_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429756_at | - | - |
Mouse Genome 430 2.0 Array | 1435024_at | - | - |
Mouse Genome 430 2.0 Array | 1440743_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1440743_at | - | - |
Mouse Genome 430 2.0 Array | 1435024_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74356_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74356_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800021602.mm.1 | NM_001166394.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021602.mm.1 | NM_001166394.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021602.mm.1 | NM_028888.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021602.mm.1 | NM_028888.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021602.mm.1 | TC0800002814.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429756_PM_at | - | - |
Mouse Expression Set 430 | 1429756_at | - | - |
Mouse Expression Set 430 | 1440743_at | - | - |
Mouse Expression Set 430 | 1435024_PM_at | - | - |
Mouse Expression Set 430 | 1435024_at | - | - |
Mouse Expression Set 430 | 1440743_PM_at | - | - |
Murine Genome U74v2 Set | 168249_PM_at | - | - |
Murine Genome U74v2 Set | 168249_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440743_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435024_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429756_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1440743_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1435024_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429756_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 469077 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 102260059 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103060121 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1237967 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244473 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638889 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2436781 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2699704 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1260404 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244473 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1231555 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237967 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2638889 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103060121 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103060121 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231555 | - | - |
