Assay Details
Gene Symbol: Nfix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081981.2 | NP_001075450.1 | 4 - 5 | 1687 | 60 | |
NM_001081982.2 | NP_001075451.1 | 4 - 5 | 1012 | 60 | ||
NM_001297601.1 | NP_001284530.1 | 4 - 5 | 1687 | 60 | ||
NM_010906.3 | NP_035036.1 | 4 - 5 | 1012 | 60 | ||
GenBank mRNA | AK158568.1 | - | 4 - 5 | 1690 | 60 | |
BC003766.1 | - | 4 - 5 | 734 | 3491917 | 60 | |
BC090626.1 | - | 4 - 5 | 714 | 60 | ||
S81451.1 | - | 3 - 4 | 698 | 60 | ||
U57636.1 | - | 3 - 4 | 689 | 60 | ||
Y07688.1 | - | 4 - 5 | 1014 | 60 | ||
Y07689.1 | - | 4 - 5 | 816 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
