Assay Details
| Gene Symbol: Nfix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081981.2 | NP_001075450.1 | 8 - 9 | 2240 | 82 | |
| GenBank mRNA | BC090626.1 | - | 8 - 9 | 1267 | 82 | |
| S81451.1 | - | 7 - 8 | 1251 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfix
Genomic Map