Assay Details
Gene Symbol: Syce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048145.1 | NP_001041610.1 | 9 - 10 | 628 | 71 | |
NM_001286494.1 | NP_001273423.1 | 8 - 9 | 565 | 71 | ||
NM_029139.1 | NP_083415.1 | 10 - 11 | 692 | 71 | ||
GenBank mRNA | AK015606.1 | - | 10 - 11 | 688 | 71 | |
AY899935.1 | - | 9 - 10 | 628 | 71 | ||
AY899936.1 | - | 10 - 11 | 692 | 71 | ||
BC100478.1 | - | 8 - 9 | 546 | 71 | ||
BC141326.1 | - | 9 - 10 | 628 | 9055938 | 71 | |
BC141347.1 | - | 9 - 10 | 628 | 9055959 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179663 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163634 | AY899935.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163634 | AY899936.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163634 | BB616021.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163634 | BC141326.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2163634 | BC141347.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025755 | AY899935.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025755 | AY899936.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025755 | NM_001048145.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025755 | NM_029139.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025755 | AK015606.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458779 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593807 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745206 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158660 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435069 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 12129 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025755 | AY899935.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025755 | AY899936.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025755 | NM_001048145.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025755 | NM_029139.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025755 | AK015606.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163634 | AY899935.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163634 | AY899936.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163634 | BB616021.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163634 | BC141326.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2163634 | BC141347.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06647295 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06647313 | - | - |
Whole Mouse Genome G4122A | A_52_P354333 | AY899936.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354333 | NM_029139.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P354333 | AK015606.1 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045118 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045110 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07045113 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26179663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179685 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179683 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179684 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179664 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179674 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179698 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179695 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179706 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26179661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179733 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179675 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179698 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179724 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07193140 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07427374 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07048433 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06675859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179732 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06804861 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07379237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179743 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07154394 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07142848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06853783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179773 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179741 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07427375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06804869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07045119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179668 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07433297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06804873 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06832669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07427371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179750 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179752 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179736 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179757 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179754 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26179714 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10575682 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575676 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575684 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575678 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575673 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575674 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575670 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575681 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575677 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575683 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575675 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575679 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10575680 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432442_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10575662 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10575672 | - | - |
Mouse Genome 430 2.0 Array | 1432442_at | - | - |
Mouse Genome 430 2.0 Array | 1432442_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 668110_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 668110_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800011155.mm.1 | NM_001048145.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011155.mm.1 | NM_001048145.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011155.mm.1 | NM_001286494.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011155.mm.1 | NM_029139.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011155.mm.1 | NM_029139.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011155.mm.1 | TC0800001357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800011154.mm.1 | NM_001048145.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011154.mm.1 | NM_001048145.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011154.mm.1 | NM_001286494.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011154.mm.1 | NM_029139.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011154.mm.1 | NM_029139.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011154.mm.1 | TC0800001357.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800011153.mm.1 | NM_001048145.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011153.mm.1 | NM_001048145.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011153.mm.1 | NM_001286494.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011153.mm.1 | NM_029139.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011153.mm.1 | NM_029139.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800011153.mm.1 | TC0800001357.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432442_PM_at | - | - |
Mouse Expression Set 430 | 1432442_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432442_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432442_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 801818 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2521283 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3065871 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3143842 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3143842 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3065871 | - | - |
