Assay Details
Gene Symbol: Syce1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048145.1 | NP_001041610.1 | 3 - 4 | 211 | 64 | |
NM_001286494.1 | NP_001273423.1 | 2 - 3 | 148 | 64 | ||
NM_029139.1 | NP_083415.1 | 3 - 4 | 211 | 64 | ||
GenBank mRNA | AK015606.1 | - | 3 - 4 | 207 | 64 | |
AY899935.1 | - | 3 - 4 | 211 | 64 | ||
AY899936.1 | - | 3 - 4 | 211 | 64 | ||
BB616021.1 | - | 3 - 4 | 191 | 64 | ||
BC100478.1 | - | 2 - 3 | 129 | 64 | ||
BC141326.1 | - | 3 - 4 | 211 | 9055938 | 64 | |
BC141347.1 | - | 3 - 4 | 211 | 9055959 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
