Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800011150.mm.1 |
NM_001048145.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011150.mm.1 |
NM_001048145.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011150.mm.1 |
NM_001286494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011150.mm.1 |
NM_029139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011150.mm.1 |
NM_029139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011150.mm.1 |
TC0800001357.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800011149.mm.1 |
NM_029139.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011149.mm.1 |
NM_029139.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800011149.mm.1 |
TC0800001357.mm.1 |
MTA TC |