Assay Details
Gene Symbol: Adgrg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033468.3 | NP_001028640.2 | 10 - 11 | 1270 | 65 | |
NM_001145972.1 | NP_001139444.1 | 10 - 11 | 1278 | 65 | ||
GenBank mRNA | AK028878.1 | - | 10 - 11 | 1271 | 65 | |
AK156147.1 | - | 10 - 11 | 1389 | 65 | ||
AK156609.1 | - | 10 - 11 | 1389 | 65 | ||
BC144851.1 | - | 10 - 11 | 1239 | 9053399 | 65 | |
BC145803.1 | - | 10 - 11 | 1239 | 40131106 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156638 | BY547428.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156638 | AK156609.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156638 | AK028878.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156638 | AK156147.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2156638 | NM_001033468.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610898 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541780 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587842 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358226 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 683794 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766535 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 334121 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 333963 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 17732 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165086 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156638 | BY547428.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156638 | AK156609.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156638 | AK028878.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156638 | AK156147.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2156638 | NM_001033468.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080288 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080295 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080283 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080298 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080282 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26080294 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06603960 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165086 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01802027 | - | - |
Whole Mouse Genome G4122A | A_52_P200686 | AK156609.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200686 | AY255569.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200686 | BC144851.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200686 | BC145803.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P200686 | AK028878.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080247 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080339 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080369 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080367 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080322 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080326 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080333 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080359 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080402 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080432 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080309 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080406 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080422 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080342 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080311 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080377 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26080324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080306 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06728986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080307 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080277 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080270 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06769658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080301 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080273 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06769660 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07150262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06769657 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06874945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080279 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080400 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06683288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06769659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06823179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07124066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07304697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07022788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26080275 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10574254 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574253 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574256 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574250 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574255 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574252 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574249 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574248 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574247 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574257 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574251 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10574258 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436230_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10574246 | - | - |
Mouse Genome 430 2.0 Array | 1436230_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1436230_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382045_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382045_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800008800.mm.1 | NM_001033468.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008800.mm.1 | NM_001033468.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008800.mm.1 | NM_001145972.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008800.mm.1 | NM_001145972.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800008800.mm.1 | TC0800001125.mm.1 | MTA TC |
Mouse Expression Set 430 | 1436230_PM_at | - | - |
Mouse Expression Set 430 | 1436230_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436230_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436230_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 865635 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1238976 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2856926 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2856926 | - | - |
