Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800008783.mm.1 |
NM_001033468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008783.mm.1 |
NM_001033468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008783.mm.1 |
NM_001145972.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008783.mm.1 |
NM_001145972.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008783.mm.1 |
TC0800001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008785.mm.1 |
NM_001033468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008785.mm.1 |
NM_001033468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008785.mm.1 |
NM_001145972.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008785.mm.1 |
NM_001145972.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008785.mm.1 |
TC0800001125.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800008782.mm.1 |
NM_001033468.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008782.mm.1 |
NM_001033468.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008782.mm.1 |
NM_001145972.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008782.mm.1 |
NM_001145972.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800008782.mm.1 |
TC0800001125.mm.1 |
MTA TC |