Assay Details
| Gene Symbol: Tcf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037877.3 | NP_001032966.1 | 12 - 13 | 1546 | 65 | |
| NM_001037878.3 | NP_001032967.1 | 12 - 13 | 1546 | 65 | ||
| NM_001286362.1 | NP_001273291.1 | 11 - 12 | 1471 | 65 | ||
| NM_001286363.1 | NP_001273292.1 | 11 - 12 | 1471 | 65 | ||
| NM_025804.3 | NP_080080.2 | 12 - 13 | 1546 | 65 | ||
| GenBank mRNA | AK003180.1 | - | 12 - 13 | 1433 | 65 | |
| AK013754.1 | - | 12 - 13 | 1235 | 65 | ||
| AK045397.1 | - | 11 - 12 | 1399 | 65 | ||
| AK050274.1 | - | 12 - 13 | 63 | |||
| AK076151.1 | - | 6 - 7 | 633 | 65 | ||
| AK088360.1 | - | 12 - 13 | 1434 | 65 | ||
| AK089964.1 | - | 12 - 13 | 1435 | 65 | ||
| AK122430.1 | - | 12 - 13 | 1401 | 65 | ||
| AK148082.1 | - | 12 - 13 | 1408 | 65 | ||
| AK150232.1 | - | 12 - 13 | 1433 | 65 | ||
| AK150918.1 | - | 12 - 13 | 1427 | 65 | ||
| AK150942.1 | - | 12 - 13 | 1420 | 65 | ||
| AK151143.1 | - | 12 - 13 | 1420 | 65 | ||
| AK152178.1 | - | 12 - 13 | 1420 | 65 | ||
| AK152494.1 | - | 12 - 13 | 1420 | 65 | ||
| AK161627.1 | - | 12 - 13 | 1422 | 65 | ||
| AK167837.1 | - | 12 - 13 | 1420 | 65 | ||
| AK168167.1 | - | 12 - 13 | 1420 | 65 | ||
| AK212702.1 | - | 2 - 3 | 65 | |||
| AK219103.1 | - | 2 - 3 | 205 | 65 | ||
| BC016130.1 | - | 12 - 13 | 1408 | 65 | ||
| BC025071.1 | - | 12 - 13 | 1407 | 3498003 | 65 | |
| BC046768.1 | - | 12 - 13 | 1504 | 5707312 | 65 | |
| EU624323.1 | - | 11 - 12 | 1309 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tcf25
Genomic Map