Assay Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163262.1 | NP_001156734.1 | 19 - 20 | 2286 | 105 | |
| NM_028941.1 | NP_083217.1 | 19 - 20 | 2109 | 105 | ||
| GenBank mRNA | AK016712.1 | - | 19 - 20 | 2110 | 105 | |
| AK165728.1 | - | 8 - 9 | 841 | 105 | ||
| AK173234.1 | - | 13 - 14 | 4536 | 105 | ||
| BC028834.1 | - | 17 - 18 | 1778 | 105 | ||
| BC158118.1 | - | 20 - 21 | 2163 | 9088191 | 105 | |
More Information
Set Membership: |
|

Cmip
Genomic Map