Assay Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008261.2 | NP_032287.2 | 3 - 4 | 501 | 63 | |
| GenBank mRNA | AK143948.1 | - | 3 - 4 | 526 | 63 | |
| BC039220.1 | - | 3 - 4 | 501 | 4238842 | 63 | |
| D29015.1 | - | 3 - 4 | 428 | 63 | ||
| EF193391.1 | - | 3 - 4 | 300 | 63 | ||
| EF193393.1 | - | 3 - 4 | 326 | 63 | ||
| FJ608819.1 | - | 4 - 5 | 455 | 63 | ||
| FJ608820.1 | - | 4 - 5 | 455 | 63 | ||
| FJ608821.1 | - | 4 - 5 | 455 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnf4a
Genomic Map