Assay Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008261.2 | NP_032287.2 | 4 - 5 | 608 | 60 | |
| GenBank mRNA | AK143948.1 | - | 4 - 5 | 633 | 60 | |
| BC039220.1 | - | 4 - 5 | 608 | 4238842 | 60 | |
| EF193391.1 | - | 4 - 5 | 407 | 60 | ||
| EF193393.1 | - | 4 - 5 | 433 | 60 | ||
| FJ608819.1 | - | 5 - 6 | 562 | 60 | ||
| FJ608820.1 | - | 5 - 6 | 562 | 60 | ||
| FJ608821.1 | - | 5 - 6 | 562 | 60 | ||
More Information
Set Membership: |
|

Hnf4a
Genomic Map