Assay Details
| Gene Symbol: Hnf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008261.2 | NP_032287.2 | 6 - 7 | 849 | 65 | |
| GenBank mRNA | AK143948.1 | - | 6 - 7 | 874 | 65 | |
| BC039220.1 | - | 6 - 7 | 849 | 4238842 | 65 | |
| D29015.1 | - | 6 - 7 | 776 | 65 | ||
| EF193391.1 | - | 6 - 7 | 648 | 65 | ||
| EF193393.1 | - | 6 - 7 | 674 | 65 | ||
| FJ608819.1 | - | 7 - 8 | 803 | 65 | ||
| FJ608820.1 | - | 7 - 8 | 803 | 65 | ||
| FJ608821.1 | - | 7 - 8 | 803 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hnf4a
Genomic Map