Assay Details
| Gene Symbol: Acd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012638.1 | NP_001012656.1 | 8 - 9 | 816 | 82 | |
| GenBank mRNA | AK032683.1 | - | 8 - 9 | 796 | 82 | |
| AK052768.1 | - | 920 | 82 | |||
| AK145959.1 | - | 8 - 9 | 913 | 82 | ||
| AK160599.1 | - | 8 - 9 | 887 | 82 | ||
| AK170606.1 | - | 7 - 8 | 797 | 82 | ||
| AK171345.1 | - | 6 - 7 | 574 | 82 | ||
| AK171458.1 | - | 8 - 9 | 806 | 82 | ||
| BC138694.1 | - | 8 - 9 | 781 | 8861716 | 82 | |
| BC138695.1 | - | 8 - 9 | 781 | 8861717 | 82 | |
| BC145384.1 | - | 8 - 9 | 882 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145959.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
telomere maintenance
skeletal system development
urogenital system development
intracellular protein transport
telomere capping
embryonic limb morphogenesis
protection from non-homologous end joining at telomere
telomere assembly
negative regulation of telomere maintenance via telomerase
segmentation
positive regulation of telomerase activity
positive regulation of single-stranded telomeric DNA binding
protein localization to chromosome, telomeric region
DNA binding
protein binding
DNA polymerase binding
telomere maintenance
skeletal system development
urogenital system development
intracellular protein transport
telomere capping
embryonic limb morphogenesis
protection from non-homologous end joining at telomere
telomere assembly
negative regulation of telomere maintenance via telomerase
segmentation
positive regulation of telomerase activity
positive regulation of single-stranded telomeric DNA binding
protein localization to chromosome, telomeric region
DNA binding
protein binding
DNA polymerase binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26135463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26135464 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26135462 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042953 | AK171458.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042953 | BC004682.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042953 | BC138694.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042953 | BC138695.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042953 | BC145384.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715387 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042953 | AK171458.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042953 | BC004682.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042953 | BC138694.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042953 | BC138695.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042953 | BC145384.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P540399 | BC004682.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135460 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135450 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135458 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135446 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135467 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135448 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135445 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26135454 | - | - |
| Whole Mouse Genome G4122A | A_52_P540399 | BC004682.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06938709 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06938717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26135463 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26135464 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26135462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26135403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07396261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135446 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07396254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06938692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135455 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26135445 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10581322 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581325 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581320 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581318 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581323 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581319 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581316 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581321 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581314 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581324 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581317 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10581315 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420882_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10581311 | - | - |
| Mouse Genome 430 2.0 Array | 1456916_at | - | - |
| Mouse Genome 430 2.0 Array | 1420882_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1420882_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1456916_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 497652_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 497652_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800021784.mm.1 | NM_001012638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021784.mm.1 | TC0800002831.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021782.mm.1 | NM_001012638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021782.mm.1 | TC0800002831.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021781.mm.1 | NM_001012638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021781.mm.1 | TC0800002831.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021783.mm.1 | NM_001012638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021783.mm.1 | TC0800002831.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800021785.mm.1 | NM_001012638.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800021785.mm.1 | TC0800002831.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420882_PM_a_at | - | - |
| Mouse Expression Set 430 | 1420882_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420882_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420882_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 103840463 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 103840463 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219304 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 103840463 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103840463 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2930819 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2930819 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1219304 | - | - |

Acd
Genomic Map