Assay Details
| Gene Symbol: Ada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272052.1 | NP_001258981.1 | 1 - 2 | 181 | 72 | |
| NM_007398.4 | NP_031424.1 | 1 - 2 | 181 | 72 | ||
| GenBank mRNA | AF483480.1 | - | 1 - 2 | 33 | 72 | |
| AF483481.1 | - | 1 - 2 | 33 | 72 | ||
| AK075899.1 | - | 1 - 2 | 120 | 72 | ||
| AK167489.1 | - | 1 - 2 | 120 | 72 | ||
| CT010224.1 | - | 1 - 2 | 33 | 72 | ||
| CT010255.1 | - | 1 - 2 | 33 | 72 | ||
| M10319.1 | - | 1 - 2 | 103 | 72 | ||
More Information
Set Membership: |
|

Ada
Genomic Map