Assay Details
| Gene Symbol: Ada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272052.1 | NP_001258981.1 | 5 - 6 | 628 | 95 | |
| NM_007398.4 | NP_031424.1 | 5 - 6 | 628 | 95 | ||
| GenBank mRNA | AF483480.1 | - | 5 - 6 | 480 | 95 | |
| AF483481.1 | - | 5 - 6 | 480 | 95 | ||
| AK075899.1 | - | 5 - 6 | 567 | 95 | ||
| AK167489.1 | - | 5 - 6 | 567 | 95 | ||
| BC002075.1 | - | 5 - 6 | 3484465 | 95 | ||
| CT010224.1 | - | 5 - 6 | 95 | |||
| CT010255.1 | - | 5 - 6 | 480 | 95 | ||
| M10319.1 | - | 5 - 6 | 550 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ada
Genomic Map