Assay Details
| Gene Symbol: A530013C23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015500.3 | 1 - 2 | 85 | 62 | ||
| GenBank mRNA | AK054128.1 | - | 1 - 2 | 55 | 62 | |
| AK162490.1 | - | 1 - 2 | 81 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

A530013C23Rik
Genomic Map