Assay Details
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153065.3 | NP_694705.2 | 4 - 5 | 482 | 139 | |
GenBank mRNA | AK134274.1 | - | 4 - 5 | 483 | 139 | |
AK138442.1 | - | 4 - 5 | 506 | 139 | ||
AK146469.1 | - | 4 - 5 | 506 | 139 | ||
BC011321.1 | - | 3 - 4 | 359 | 4007058 | 139 | |
BC024730.1 | - | 4 - 5 | 482 | 139 | ||
BC024981.1 | - | 4 - 5 | 482 | 139 | ||
BC033398.1 | - | 4 - 5 | 482 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC011321.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
