Assay Details
| Gene Symbol: 1500012F01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081005.1 | NP_001074474.1 | 2 - 3 | 456 | 76 | |
| GenBank mRNA | AI847202.1 | - | 3 - 4 | 76 | ||
| AK005231.1 | - | 2 - 3 | 169 | 76 | ||
| BC042795.1 | - | 2 - 3 | 125 | 76 | ||
| BF467288.1 | - | 2 - 3 | 456 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005231.1, NM_001081005.1 transcripts.
Set Membership: |
|

1500012F01Rik
Genomic Map