Assay Details
| Gene Symbol: Stau1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109905.2 | NP_001103375.1 | 5 - 6 | 880 | 107 | |
| NM_001109906.2 | NP_001103376.1 | 5 - 6 | 898 | 107 | ||
| NM_001291160.1 | NP_001278089.1 | 5 - 6 | 880 | 107 | ||
| NR_111895.1 | 4 - 5 | 632 | 107 | |||
| GenBank mRNA | AF395842.1 | - | 4 - 5 | 601 | 107 | |
| AK005002.2 | - | 5 - 6 | 821 | 107 | ||
| AK139532.1 | - | 5 - 6 | 827 | 107 | ||
| AK145826.1 | - | 5 - 6 | 865 | 107 | ||
| AK151835.1 | - | 5 - 6 | 686 | 107 | ||
| AK152269.1 | - | 5 - 6 | 692 | 107 | ||
| AK152744.1 | - | 5 - 6 | 686 | 107 | ||
| AK158952.1 | - | 1 - 2 | 482 | 107 | ||
| AK159602.1 | - | 5 - 6 | 815 | 107 | ||
| AK159781.1 | - | 5 - 6 | 850 | 107 | ||
| AK160979.1 | - | 5 - 6 | 839 | 107 | ||
| AK167637.1 | - | 5 - 6 | 821 | 107 | ||
| AY391773.1 | - | 4 - 5 | 631 | 107 | ||
| BC012959.1 | - | 5 - 6 | 844 | 4188394 | 107 | |
| BC082277.1 | - | 5 - 6 | 817 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stau1
Genomic Map