Assay Details
| Gene Symbol: Rnf114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030743.5 | NP_109668.2 | 5 - 6 | 1279 | 63 | |
| GenBank mRNA | AF282919.2 | - | 3 - 4 | 423 | 63 | |
| AF502145.1 | - | 3 - 4 | 426 | 63 | ||
| AK079751.1 | - | 3 - 4 | 433 | 63 | ||
| AK088441.1 | - | 3 - 4 | 433 | 63 | ||
| AK139030.1 | - | 3 - 4 | 413 | 63 | ||
| AK148290.1 | - | 3 - 4 | 440 | 63 | ||
| AK148392.1 | - | 3 - 4 | 428 | 63 | ||
| AK150086.1 | - | 3 - 4 | 420 | 63 | ||
| AK150263.1 | - | 3 - 4 | 420 | 63 | ||
| AK150700.1 | - | 2 - 3 | 156 | 63 | ||
| AK152687.1 | - | 3 - 4 | 420 | 63 | ||
| AK153452.1 | - | 3 - 4 | 410 | 63 | ||
| AK159818.1 | - | 3 - 4 | 410 | 63 | ||
| AK163062.1 | - | 2 - 3 | 266 | 63 | ||
| AK194838.1 | - | 1 - 2 | 78 | 63 | ||
| BC023125.1 | - | 3 - 4 | 438 | 63 | ||
| BC043446.1 | - | 3 - 4 | 405 | 63 | ||
| BC054416.1 | - | 3 - 4 | 415 | 3672897 | 63 | |
| BC085146.1 | - | 5 - 6 | 1279 | 6812845 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf114
Genomic Map