Assay Details
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008707.3 | NP_032733.1 | 2 - 3 | 247 | 83 | |
GenBank mRNA | AF043326.1 | - | 2 - 3 | 259 | 83 | |
AK146523.1 | - | 2 - 3 | 262 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
