Assay Details
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008707.3 | NP_032733.1 | 8 - 9 | 999 | 56 | |
GenBank mRNA | AF043326.1 | - | 8 - 9 | 1011 | 56 | |
AK146523.1 | - | 8 - 9 | 1014 | 56 | ||
BC016526.1 | - | 8 - 9 | 999 | 3589678 | 56 | |
BC021635.1 | - | 8 - 9 | 1013 | 3989540 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
