Assay Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152813.3 | NP_690026.2 | 7 - 8 | 1371 | 101 | |
| GenBank mRNA | AK011892.1 | - | 3 - 4 | 303 | 101 | |
| AK046669.1 | - | 7 - 8 | 1337 | 101 | ||
| AK144950.1 | - | 2444 | 101 | |||
| AK164809.1 | - | 6 - 7 | 1613 | 101 | ||
| AK166977.1 | - | 5 - 6 | 2712 | 101 | ||
| AK173311.1 | - | 6 - 7 | 1619 | 101 | ||
| BC031392.1 | - | 7 - 8 | 1315 | 4952407 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plcd3
Genomic Map