Assay Details
| Gene Symbol: Acbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025988.2 | NP_080264.1 | 3 - 4 | 632 | 73 | |
| GenBank mRNA | AK148438.1 | - | 127 | 73 | ||
| BC048371.1 | - | 2 - 3 | 546 | 6396076 | 73 | |
| BI734653.1 | - | 2 - 3 | 171 | 73 | ||
| DN173459.1 | - | 3 - 4 | 589 | 73 | ||
More Information
Set Membership: |
|

Acbd4
Genomic Map