Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100016079.mm.1 |
NM_025988.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016079.mm.1 |
NM_025988.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016079.mm.1 |
TC1100001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016092.mm.1 |
NM_025988.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016092.mm.1 |
NM_025988.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016092.mm.1 |
TC1100001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016098.mm.1 |
NM_025988.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016098.mm.1 |
NM_025988.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016098.mm.1 |
TC1100001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016081.mm.1 |
NM_025988.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016081.mm.1 |
NM_025988.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016081.mm.1 |
TC1100001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016094.mm.1 |
NM_025988.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016094.mm.1 |
NM_025988.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016094.mm.1 |
TC1100001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100016077.mm.1 |
NM_025988.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016077.mm.1 |
NM_025988.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100016077.mm.1 |
TC1100001705.mm.1 |
MTA TC |