Assay Details
| Gene Symbol: Pitpnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024927.2 | NP_001020098.1 | 2 - 3 | 334 | 78 | |
| NM_001081641.1 | NP_001075110.1 | 2 - 3 | 334 | 78 | ||
| GenBank mRNA | AK147444.1 | - | 2 - 3 | 291 | 78 | |
| AK147630.1 | - | 2 - 3 | 167 | 78 | ||
| BC112411.1 | - | 1 - 2 | 709 | 40061037 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnm3
Genomic Map