Assay Details
| Gene Symbol: Elp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081381.1 | NP_001074850.1 | 5 - 6 | 626 | 89 | |
| GenBank mRNA | AK011270.1 | - | 6 - 7 | 782 | 89 | |
| AK075986.1 | - | 5 - 6 | 623 | 89 | ||
| BC048732.1 | - | 5 - 6 | 601 | 6512688 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Elp6
Genomic Map